ART® 1000E Self-Sealing Barrier Pipette Tips, Molecular BioProducts
Supplier: Thermo Scientific
2079EIW 2079E
53225-782CS 454.62 CAD
53225-782 72830-042
ART® 1000E Self-Sealing Barrier Pipette Tips, Molecular BioProducts
Pipette Tips
Tips ensure 100% protection against aerosol and liquid contamination.
  • Pre-sterilized tips
  • Tips provide an ideal fit for most 1000 µl pipettors
The ART® barrier traps contaminants, keeping them out of pipettors and samples. This also protects the sensitive inner components of the pipettor, prolonging pipettor life. ART® tips are ideal for nucleic amplification procedures, pipetting of radioactive samples, or any application where critical sample handling is required.
Certifications: Certified RNase-, DNase-, DNA-, and pyrogen-free.
Packaging: More Product Information