ART® 300 Barrier Pipette Tips, Sterile, Thermo Scientific
Supplier: Thermo Scientific ART
ART®
2070 2070-HR 2070-05-HR
53509-508CS 1811.09 CAD
53509-508 89413-264 89413-268
ART® 300 Barrier Pipette Tips, Sterile, Thermo Scientific
Pipette Tips
Tips ensure 100% protection against aerosol and liquid contamination.
  • Pre-sterilized tips
  • Secure fit and extra space for repeat pipetting
The ART® barrier traps contaminants, keeping them out of pipettors and samples. This also protects the sensitive inner components of the pipettor, prolonging pipettor life. ART® tips are ideal for nucleic amplification procedures, pipetting of radioactive samples, or any application where critical sample handling is required. Tips are compatible with Eppendorf®, Transferpette®, Finnpipette®, Biohit®, and Rainin® series pipettors.
Certifications: Certified RNase-, DNase-, DNA-, and pyrogen-free.
Ordering information: Available in both traditional lift-off and versatile hinged racks.
Packaging: Sterile tips are packaged 96/tray, 10 trays per pack.