Eppendorf twin.tec® PCR Plates 384-Well, 45 µl
Supplier: Eppendorf
951020702 951020711 951020729 951020737 951020745 0030129342 0030129350 951020573 951020594 951020552 951020539 951020541 0030129628 0030113578
CA12000-658PK 546.58 CAD
CA12000-658 CA12000-660 CA12000-662 CA12000-664 CA12000-666 CA89136-710 CA89136-712 CA95041-420 CA95041-422 CA95041-424 CA95041-426 CA95041-428 76019-008 CA77395-530
Eppendorf twin.tec® PCR Plates 384-Well, 45 µl
Microplates PCR Plates
Eppendorf's twin.tec® 384-well, 45 µl PCR Plates feature a polycarbonate frame and thin-walled polypropylene wells ensuring optimal heat transfer and high rigidity. With raised well rims, these plates enable an effective seal, minimizing the risk of cross-contamination and contributing to consistently reliable results.
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample 
  • Exceptionally solid and torque-resistant
More Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification Renewable Energy Certificate, ISO 14001