AMPLATE™ Thin-Walled PCR Plates, Simport Scientific
Supplier: Simport
Amplate™
T323-48G T323-96B T323-24N T323-24Y T323-24B T323-48Y T323-48R T323-96R T323-24N T323-96G T323-48B T323-24R T323-96N T323-96R T323-24B T323-96Y T323-48R T323-24G T323-96N T323-48N T323-24Y T323-96B T323-24R T323-48N T323-48B T323-48G T323-24G T323-96Y T323-96G
CA48648-510CS 95.61 CAD
CA48648-510 CA60872-670 89429-038 60872-660 89429-036 89429-042 89429-040 CA60872-662 89429-046 60872-656 CA48648-509 CA48648-508 89429-044 60872-655 CA48648-507 CA60872-660 89429-048 89429-052 CA60872-656 89429-050 89429-034 CA60872-652 89429-056 89429-054 CA48648-515 CA48648-514 CA48648-513 CA48648-512 CA48648-511
AMPLATE™ Thin-Walled PCR Plates, Simport Scientific
Microplates PCR Plates
These PCR plates are thin-walled and designed for rapid thermal transfer
The flexible design of plates allows them to be easily cut into sections of 24, 32, or 48 tubes. They accommodate differences in expansion coefficients between the metal thermal cycler block and polypropylene tubes. An alphanumeric grid helps with sample identification, and a corner is cut away to facilitate orientation of the plate. For easy sealing, cylindrical walls do not extend above the plates. Plates are certified RNase-, DNase-, and DNA-free.
Plates are precision-molded to ensure well-to-well and plate-to-plate uniformity. The insides of the tubes are smooth and have an inert surface on which enzymes and nucleic acids do not bind. Each well has a capacity of 0.2mL.
Accessories information: The domed 8-Strip Cap is designed to fit the T323-96 PCR Plate series.
Ordering information: Packed in tamperproof resealable bags of 10 plates.