µltraAmp™ 96-Well Semi-Skirted PCR Plates, Sorenson BioScience
Supplier: Sorenson Bioscience
35800 35810 35820 35830 35840 35850 37380 37390
14216-140CS 443.17 CAD
14216-140 14216-142 14216-144 14216-146 14216-148 14230-056 14230-088 14230-090
µltraAmp™ 96-Well Semi-Skirted PCR Plates, Sorenson BioScience
Microplates PCR Plates
Skirt allows robotic handling and sample labeling
Certified nonpyrogenic and free of RNase, DNase, human DNA, and PCR inhibitors.
Raised rim around perimeter for aligning purification plates. For use with Applied Biosystems thermal cyclers and DNA analyzers. Precision-designed molds and tightly controlled processes create high-quality plates that exhibit minimal warp before and after thermal cycling. Bold alphanumeric grid. Black and white plates are designed for Real-Time PCR and other fluorescence based assays. Black plates eliminate background signal and well-to-well crosstalk. White plates deliver maximum signal amplification.