ART® 100E Self-Sealing Barrier Pipette Tips, Molecular BioProducts
Supplier: Thermo Scientific ART
ART®
2065E
72830-032CS 1742.83 CAD
72830-032
ART® 100E Self-Sealing Barrier Pipette Tips, Molecular BioProducts
Pipette Tips
Tips ensure 100% protection against aerosol and liquid contamination.
  • Pre-sterilized tips
  • Universal design for a variety of 100 µl pipettors
The ART® barrier traps contaminants, keeping them out of pipettors and samples. This also protects the sensitive inner components of the pipettor, prolonging pipettor life. ART® tips are ideal for nucleic amplification procedures, pipetting of radioactive samples, or any application where critical sample handling is required.
Certifications: Certified RNase-, DNase-, DNA-, and pyrogen-free.
Packaging: Sterile tips are packaged 96/tray, 10 trays per pack. Non sterile tips are bulk packed.