Axygen® Polypropylene PCR Plates, Corning
Supplier: Corning
PCR-24-C PCR-32-C PCR-48-C
10011-164CS 285.82 CAD
10011-164 10011-180 10011-205
Axygen® Polypropylene PCR Plates, Corning
Microplates PCR Plates
PP. Designed to fit 96-well thermal cycler blocks.
  • Ultra-thin walls of uniform thickness accelerate heat transfer, leading to shorter cycle times and more consistent results
  • Certified free from DNase, RNase and endotoxins for highest sample integrity
These multiwell plates are specifically designed for PCR applications and are ideal for thermal transfer application. Max. RCF 4000 g.