Pipette Tips for Rainin LiteTouch™ Pipettors, Molecular BioProducts
Supplier: Thermo Scientific
3752 2749 3721 2769 3751 3782 3781 2779 2749-05 2769-05 2779-05 2789
72830-268CS 1860.94 CAD
72830-268 72830-270 72830-272 72830-276 72830-278 72830-280 72830-282 72830-284 72830-444 72830-454 72830-471 82031-392
Pipette Tips for Rainin LiteTouch™ Pipettors, Molecular BioProducts
Pipette Tips
ART® self-sealing barriers trap contaminants, such as aerosols and liquids, keeping them out of pipettors and samples.
  • ART® tips are presterilized
  • Several tips are available with low retention technology
Tips are available in non sterile packaging, in sterile packaging, or with ART self-sealing barrier.
This allows the user to conduct PCR procedures and pipette radioactive or infectious samples. This permits more accurate sample dispensing, an increased level of reproducibility, and reduced sample loss due to increased adhesion to pipette tips.
Certifications: Certified RNase-, DNase-, DNA-, and pyrogen-free.
Packaging: All tips are packaged 96/rack.