MµltiGuard³™ Universal Fit Pipette Tips, Sterile, Sorenson BioScience
Supplier: Sorenson Bioscience
17370 17370T
53550-450PK 444.87 CAD
53550-450 14229-980
MµltiGuard³™ Universal Fit Pipette Tips, Sterile, Sorenson BioScience
Pipette Tips
Tips can be used with 20, 100, and 200 µl pipettors and accurately deliver 1 to 200 µl.
  • Tip 14229-980 exhibits low binding properties
They feature a unique design and filter placement that allow precise volume delivery with most popular brand pipettors.
Certifications: Certified RNase- and DNase-free.
Ordering information: Supplied in polypropylene racks of 96 tips.