KEMRESIN® Countertops, Kewaunee®
Supplier: Kewaunee
KEMRESIN®
KS-R0496-00
KS-R0496-00EA 1056.42 CAD
KS-R0496-00
KEMRESIN® Countertops, Kewaunee®
Furniture Casework
Molded epoxy resin construction provides excellent physical-, chemical-, and stain-resistance.
  • Countertops are 2.5 cm (1") thick
It comes with three finished edges, including a beveled front edge; drip groove on the underside; and an attractive, black, nonglare surface.
Ordering information: Work surfaces are supplied flat or with an optional applied curb, 10.2H×1.9 cm thick (4×¾"). Please specify when ordering.