RNase AWAY® and DNA AWAY™ Surface Decontaminants, Molecular BioProducts
Supplier: Thermo Scientific
7002 7003 7005-11 7000
17810-491CS 964.95 CAD
17810-491 17810-494 53225-514 72830-022
RNase AWAY® and DNA AWAY™ Surface Decontaminants, Molecular BioProducts
Cleaning Agents Decontaminants
RNase AWAY® eliminates RNase and DNA from laboratory surfaces
DNA AWAY™ decontaminant is ideal for removing DNase and DNA contamination.
Note: Should not be used as a reagent.
The surface decontaminant is ideal for decontaminating apparatus, benchtops, glassware, and plastic ware. The decontaminant leaves no residue to interfere with gel polymerization or staining and contains no strong acids