GN Metricel® Membrane Disc Filters, Cytiva
Supplier: Cytiva
GN Metricel®
63068 66265 64677 66191ME 63069 63020 63020 66278 66191ME 64678 66191 66276 63068 66179 64679 66068 64191 66278 64679 66265 66191 64677 66539 63069 64191 66068 60016 66276 60016 66179 64678 66539
CA28148-399PK 365 CAD
CA28148-399 28148-642 28148-562 28143-326 28148-584 28148-926 CA28148-813 28148-325 CA28148-675 28148-303 87003-868 CA28148-926 CA28148-391 CA28148-290 CA77528-942 28148-675 CA28148-562 28148-399 CA28148-584 CA28143-326 28148-815 28148-813 CA28148-303 CA28148-325 CA28148-642 87003-866 CA87003-866 CA87003-868 CA28148-815 28148-391 28148-290 77528-942
GN Metricel® Membrane Disc Filters, Cytiva
Filters Membrane Filters
The GN-6 membrane is certified for the microbiological analysis of potable, waste, process, and natural waters in accordance with the Membrane Filter Technique referenced in Standard Methods for the Examination of Water and Wastewater.
  • For microbiological analysis
  • Mixed cellulose ethers
  • Easy bacterial quantification
Ideal for isolation and enumeration of total and fecal coliforms, E.
GN-6 and GN-4 Metricel® membrane filters are made of mixed cellulose esters. GN-6 grids have a unique dot grid pattern that provides easy quantification of bacterial colonies without growth inhibition or enhancement. GN-4 membranes dissolve completely using standard digestion procedures, and clear completely for minimal interference in fiber counting.
E. coli, fecal Streptococcus, fungi, and other heterotrophic bacteria. The GN-4 membrane is suitable for air monitoring applications and meets NIOSH requirements for airborne metals and asbestos monitoring.
Certifications: Meets NIOSH requirements for airborne metals and asbestos monitoring.
Ordering information: GN-6 is available gamma irradiated, individually packaged (except filter model 60016) or nonsterile, bulk packed. GN-4 is available in nonsterile bulk packs or cavities, with or without support pads.