Corning® Treated Culture Dishes, Corning
Supplier: Corning
430165 430166 430196 430167 430293 430599 431110 3261 3296 3294 3295 4615
25382-348CS 747.1 CAD
25382-348 25382-381 25382-392 25382-428 25382-439 25382-442 29186-489 33500-730 89089-840 89089-842 89089-844 76441-542
Corning® Treated Culture Dishes, Corning
Culture Dishes Cell Culture Dishes
Optically clear PS. Available with a choice of treated surface. Corning® CellBIND® surface increases surface wetability for more even and consistent cell attachment. Ultra-Low Attachment dishes feature a covalently bound hydrogel layer that minimises cell attachment, protein absorption and cellular activation.
  • With stacking beads to aid handling
  • Supplied with vents to provide consistent gas exchange
  • Sterilised by gamma radiation
  • Certified non-pyrogenic
These disposable petri-style dishes are manufactured from optically clear virgin polystyrene. Dishes are equipped with stacking beads to aid in handling and feature vents to ensure consistent gas exchange. Sterilized by gamma irradiation.
The surface minimizes cell attachment, protein absorption, and enzyme activation. It is ideal for culturing stem cells, as it promotes ebryoid body formation from ES cells. Surfaces are stable, noncytotoxic, biologically inert, and nondegradable.