polyproLABS® Base Cabinets, Air Control
Supplier: Air Control
AC-160-1830 AC-160-2430 AC-160-3030 AC-160-3630 AC-160-1836 AC-160-2436 AC-160-3036 AC-160-3636 AC-165-1830 AC-165-2430 AC-165-3030 AC-165-3630 AC-165-1836 AC-165-2436 AC-165-3036 AC-165-3636 AC-170-1830 AC-170-2430 AC-170-3030 AC-170-3630 AC-170-1836 AC-170-2436 AC-170-3036 AC-170-3636 AC-175-1830 AC-175-2430 AC-175-3030 AC-175-3630 AC-175-1836 AC-175-2436 AC-175-3036 AC-175-3636 AC-180-3630 AC-180-4230 AC-180-4830 AC-180-3636 AC-180-4236 AC-180-4836 AC-185-3630 AC-185-4230 AC-185-4830 AC-185-3636 AC-185-4236 AC-185-4836 AC-190-3630 AC-190-4230 AC-190-4830 AC-190-3636 AC-190-4236 AC-190-4836 AC-195-3630 AC-195-4230 AC-195-4830 AC-195-3636 AC-195-4236 AC-195-4836 AC-150-1830 AC-150-2430 AC-150-3030 AC-150-1836 AC-150-2436 AC-150-3036 AC-155-2436 AC-155-3630 AC-155-4230 AC-155-4830 AC-155-3636 AC-155-4236 AC-155-4836 AC-120-3030 AC-120-3630 AC-120-4230 AC-120-4830 AC-120-3036 AC-120-3636 AC-120-4236 AC-120-4836 AC-115-3030 AC-115-3630 AC-115-4230 AC-115-4830 AC-115-3036 AC-115-3636 AC-115-4236 AC-115-4836
89008-376EA 5627.5 CAD
89008-376 89008-378 89008-380 89008-382 89008-384 89008-386 89008-388 89008-390 89008-392 89008-394 89008-396 89008-398 89008-400 89008-402 89008-404 89008-406 89008-408 89008-410 89008-412 89008-414 89008-416 89008-418 89008-420 89008-422 89008-424 89008-426 89008-428 89008-430 89008-432 89008-434 89008-436 89008-438 89008-440 89008-442 89008-444 89008-446 89008-448 89008-450 89008-452 89008-454 89008-456 89008-458 89008-460 89008-462 89008-464 89008-466 89008-468 89008-470 89008-472 89008-474 89008-476 89008-478 89008-480 89008-482 89008-484 89008-486 89008-488 89008-490 89008-492 89008-494 89008-496 89008-498 89008-500 89008-502 89008-504 89008-506 89008-508 89008-510 89008-512 89008-684 89008-686 89008-688 89008-690 89008-692 89008-694 89008-696 89008-698 89008-762 89008-764 89008-766 89008-768 89008-770 89008-772 89008-774 89008-776
polyproLABS® Base Cabinets, Air Control
Furniture Casework
Corrosion-resistant casework cabinets are constructed of stress-relieved, acid-resistant, 1.3 cm (½") thick polypropylene.
  • Drawers slide quietly and evenly on a polypropylene notched groove system
Drawer panels are 1 cm (⅜") thick and feature built-in stops to prevent accidental removal. The drawer suspension system is rated to withstand numerous cycles at 45.4 kg (100 lbs.). Doors are solid 1 cm (⅜") thick with beveled edges.
Two models of cabinets include adjustable shelves made from solid 1.3 cm (½") thick polypropylene. Cabinets with two half-More Product Information