Pressure Reaction Flasks, Unjacketed, Ace Glass Incorporated
Supplier: Ace Glass
6423-05 6423-07 6423-10 6423-20 6423-30 6423-35
89054-250EA 530.87 CAD
89054-250 89054-252 89054-254 89054-256 89054-282 89054-284
Pressure Reaction Flasks, Unjacketed, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Flask Shape
These rugged, heavy-wall, round-bottom flasks are for use with the two-piece Pressure Reactor (head is sold separately) and can be used under elevated pressures
Schott-type flange is supplied with a shallow O-ring groove for use with mating grooved head.
Ordering information: Supplied with CAPFE O-ring. For head, heating mantle, silicone O-rings, or other parts, contact your Avantor representative.