Specialty Cell Culture Media

Supplier: Custom Comfort

GA1972 04-380Q 04-418Q 04-448Q BELN12-766Q
CA12001-002EA 12737.63 CAD
CA12001-002 CA12001-340 CA12001-610 CA12001-628 CA12001-772 CA12001-984 CA12001-988 CA12001-990 CA12002-026 CA12002-058 CA12002-178 CA12002-186 CA12002-188 CA12001-618 CA77578-272
Specialty Cell Culture Media
Cell Culture Media

Serum-free media provide better control over the physiological condition of cultures while allowing simplified purification and downstream processing protocols


  • Superior media for most hematopoietic cells types including dendritic cells, lymphocytes, monocytes, macrophages and granulocytes.
  • Formulations only contain human derived components
  • Highly cited in scientific literature
  • Clinical proven formulations providing process safety and constancy
  • High productivity resulting in a high viable cell yield
  • Serum Free media result in a more defined process and result in limited immune response
  • Efficient transgene expression
  • Desirable T-cell phenotype


These specialty formulations must satisfy a number of nutritional and physical requirements of cells that are normally addressed by the presence of serum. Most serum-free media contain transferrin to deliver iron and insulin to regulate the uptake of glucose.


Applications:
Proliferation of peripheral blood lymphocytes.
Proliferation of tumor infiltrating lymphocytes.
Cryopreservation of human tissue.
Cultivation of human monocytes and macrophages.
Cultivation of stem cells.
Cultivation of dendritic cells.
Cultivation of CAR-T cells.
Protein production.
Recombinant protein production.
Use of Baculovirus vector expression system (BEVS).
Versatile Medium for shaker flask or stationary culture of Sf9, Sf21, High Five® and drosophila cells.


Ordering information: Store all media at 2 to 8°C (36 to 46°F), except for UltraCHO™ Supplement 12002-028, which should be stored at –20 to –10°C (4 to 14°F).


Delivery information: Content: 1 L bottle.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR