InLab® Expert Pro 3-in-1 pH Electrodes, Double Junction, METTLER TOLEDO®
Supplier: Mettler Toledo
51343101 51343103 51343104 51343105 51343102
CA97001-610EA 948.17 CAD
CA97001-610 CA97001-612 CA97001-614 CA97001-616 CA97001-618
InLab® Expert Pro 3-in-1 pH Electrodes, Double Junction, METTLER TOLEDO®
Electrodes pH Electrodes
Electrodes are ideal for a wide range of applications, including emulsions, suspensions, effluent water, and solutions containing proteins and sulphides.
  • Integrated temperature probe
  • Solid polymeric electrolyte
  • No junction required
  • ARGENTHAL™ lead-off and silver ion trap
  • PEEK™ shaft
  • High chemical resistance
They feature an integrated temperature probe, a solid polymeric electrolyte that requires no junction; an ARGENTHAL™ lead-off and silver ion trap for optimum silver ion-free accuracy at various sample temperatures; and a robust high-grade PEEK™ shaft for high chemical resistance. Electrodes with MultiPin™ connectors feature a low-resistance, universal glass membrane. Diameter: 12 mm. Lead length: 1.2 m, except for 97001-618 which is 2 m.