InLab® Routine Pro 3-in-1 pH Electrodes, METTLER TOLEDO®
Supplier: Mettler Toledo
51343054 51343056
CA97001-596EA 860.57 CAD
CA97001-596 CA97001-600
InLab® Routine Pro 3-in-1 pH Electrodes, METTLER TOLEDO®
Electrodes pH Electrodes
General-purpose electrodes feature integrated temperature probe and ARGENTHAL™ lead-off and silver ion trap for optimum silver ion-free accuracy at various sample temperatures
  • ARGENTHAL™ lead-off and silver ion trap
  • High alkali glass body
  • Resistant to corrosives
High alkali glass body is resistant to organic or corrosive samples. Supplied with 25mL electrolyte vial. Diameter: 12mm.