Cushion-Grade Conductive Foam, Protektive Pak®
Supplier: Protektive Pak
37681 37680 37682
30000-206EA 449.48 CAD
30000-206 30000-212 30000-218
Cushion-Grade Conductive Foam, Protektive Pak®
Mailers and Shippers
Non-contaminating, non-corrosive, and non-sloughing cushioning protects items from physical shock.
  • Conductive crosslink polyethylene foam will not particulate, making it ideal for critical environments.
Resistivity range: 103 - 104 ohms/sq.
Density: 44.9 kg/m3 (2.8 lbs./cu.ft.)
Order Now
SPECIFICATIONS
Environmentally Preferable