HBIO ECM 2001 Plug Connection Cables, Red and Black, 10 ft, BTX
Supplier: Harvard Apparatus
45-0216 45-0217
82019-192EA 656.71 CAD
82019-192 82019-194
HBIO ECM 2001 Plug Connection Cables, Red and Black, 10 ft, BTX
Cables
Our cables and adapters can be used for a variety of products.
  • For use with microslides, genetrodes and tissue petri dish, ECM 2001
  • Micrograbber to Banana cables
  • Banana to Banana cables
Used to connect BTX electrofusion/electroporation systems to the electrodes and chambers used for a particular application.
Cables and adapters are needed to connect an electrofusion/electroporation system to the electrodes and chambers to be used for a particular application. For more details, please see the electrode type of interest.