Microslides Electrodes, BTX®
Supplier: Harvard Apparatus
45-0106 45-0103 45-0104 45-0105
82020-820EA 557.96 CAD
82020-820 58017-994 58017-996 58017-998
Microslides Electrodes, BTX®
Electrodes Electroporation Electrodes
Ideal for embryo manipulation, nuclear transfer, hybridoma production, and cell or protoplast fusion
Stainless steel electrodes are mounted on glass slides designed to easily fit on a microscope to allow observation of the dimer formation during electrofusion.
Ordering information: Connection to an electroporator requires banana plug-to-banana plug connector cable (82019-194).