Falcon® 96-Well Cell Culture Plates, Corning
Supplier: Corning
Falcon®
353916 353219 353075 353377 353910 351172 353910 353377 353296 353916 353072 353077 353376 353936 353227 353936 351172 353075 351177 353376 353077 353072 353296 351177 353219 353227
CA62406-015CS 288.41 CAD
CA62406-015 62406-121 80086-578 25382-208 CA15705-066 CA15705-064 CA62406-117 25382-200 21100-006 CABD353219 CA80086-578 CA11008-408 CA62406-081 89218-098 62406-015 89218-096 CA25382-208 CA62406-121 62406-117 15705-066 CA25382-200 15705-064 CA11008-410 CA21100-006 89022-352 62406-081
Falcon® 96-Well Cell Culture Plates, Corning
Microplates Cell Culture Plates
Falcon® 96-well microplates are available in a variety of configurations and surface chemistries to meet a wide range of applications with fluorescence, luminescence, radiometric or colorimetric endpoints.
  • Flat bottom, with lid
  • Black wells with clear bottom
  • Working volume: 25 to 340 µl
  • Convenient, peel-open packaging
  • Available with untreated or tissue culture-treated surface chemistries
  • Individual and Ready-Stack (RS) trays are PET (Code 1) and recyclable
Corning® Primaria™ tissue culture treatment additionally incorporates nitrogen-containing functional groups that have been shown to improve attachment and spreading of some cell types.
Falcon 96, 384, and 1536 well microplates are compatible with automation systems and meet standard ANSI/SBS footprint dimensions for microplates.