Lab Mailers and Mailing Sleeves for Specimen Transport, Sonoco ThermoSafe
Supplier: Sonoco Thermosafe
442 409 365 373 440KD 402 404 369 395 356 407 362 357 340 364 408 339 341 403 435 436 443 312 440 335 336 400 444 601 602
15713-520CS 289.85 CAD
15713-520 15714-463 15714-505 15714-525 15715-140 15715-160 16465-254 35751-310 48429-305 73320-046 73320-054 73320-058 73320-062 73320-066 73320-068 73320-070 73320-072 73320-074 73320-076 73320-080 73320-084 73320-086 82031-416 89049-968 89049-972 89049-974 89050-632 15713-507 15713-610 16191-180 16191-185 73320-036 73320-040 87006-048 89496-024 89496-026
Lab Mailers and Mailing Sleeves for Specimen Transport, Sonoco ThermoSafe
Mailers and Shippers
Lightweight, economical EPS shippers provide maximum breakage protection for known non-hazardous materials during specimen transport.
  • For VacuTainer®, Microtainer®, and other collection/transport tubes
  • Available in multi-tube, slide, and tube/slide combinations
  • Lightweight polystyrene foam saves postage
  • Reusable and recyclable
A unique array of specimen shippers fills the need for protection of frozen, refrigerated, and ambient specimens required for compliance, analysis, testing, research and development, public health, industrial hygiene and environmental use.