Infectious Shippers, Sonoco ThermoSafe
Supplier: Sonoco Thermosafe
INF3000 670
15714-540CS 1107.26 CAD
15714-540 89050-568 89496-022
Infectious Shippers, Sonoco ThermoSafe
Mailers and Shippers
ThermoSafe® infectious mailers include components required for transporting infectious substances.
  • Pre-assembled packaging
  • Includes all required components
  • Meet DOT and IATA requirements
Pre-assembled packaging includes everything for shipping infectious agents in accordance with DOT Regulation 49 CFR Parts 171 to 178 and IATA Packing Instructions 602. ThermoSafe® Infectious Shipper, part 630, includes EPS foam container, flexible foam insert, clear plastic press-lock bag, secondary packaging, absorbent material for full capacity of blood, corrugated carton and step-by-step packing instructions. Infecon® shippers include pressure vessel with stop-lock lid, compartmentalized bubble pouch, 50 mL absorbent sheet, corrugated outer carton with required markings, shipper's declarations and easy-to-follow instructions. Dry Ice Overpack protects frozen samples for over four days. The EPS foam container and lid accomodates model 630, plus 25 lb [11kg] of dry ice. Foam insert holds in place while dry ice sublimates. Included one Class 6 Infectious Substance label and one Class 9 Miscellaneous Dangerous Goods label (for dry ice), and shipping carton
Certifications: Shippers meet U.S. Department of Transportation (DOT) and IATA requirements for transporting infectious substances.
Ordering information: Compliance with UN/DOT regulations governing shipments of infectious substances in these approved shippers is contingent on the use of all supplied components. The shipper is ultimately responsible for determining the suitability of packaging for use with contents. Consult appropriate regulations.