ShipSafe® Bottle Shippers, Sonoco ThermoSafe
Supplier: Sonoco Thermosafe
347UPS 358UPS 338UPS 393UPS 743 359UPS 361UPS
14100-404CS 215.02 CAD
14100-404 14100-408 14100-416 14100-426 82031-424 14100-422 14100-424
ShipSafe® Bottle Shippers, Sonoco ThermoSafe
Mailers and Shippers
Lightweight, economical shippers are ideal for shipping reagents, pharmaceuticals, biological preparations, and other non-hazardous liquids in Boston round or regular bottles.
  • Ideal for shipping non-hazardous liquids
  • Two-piece molded EPS
  • No outer carton required
  • Reusable and recyclable
Shippers are made of two-piece molded, expanded polystyrene (EPS) foam with interlocking edges and molded-in tape grooves. No outer shipping carton is required; pressure-sensitive labels adhere directly to shipper. Shippers are reusable, recyclable, and laboratory tested and accepted for use by UPS, Parcel Post, and common carriers.