Axygen® PCR Tube Strips and Dome Cap Strips, Corning
Supplier: Corning
PCR-0208-A PCR-0208-G PCR-0208-Y PCR-02CP-A PCR-0208-CP-C PCR-02CP-C-100 PCR-0208-CP-C-100 PCR-0208-C-100
10011-760CS 3771.14 CAD
10011-760 10011-766 10011-774 10011-782 12000-206 CA10169-886 CA10169-916 CA10169-918
Axygen® PCR Tube Strips and Dome Cap Strips, Corning
Tubes PCR Tubes
These polypropylene tube strips and cap strips are specifically designed for PCR use and are ideal for thermal transfer applications.
  • Made from highly polished molds
  • Free of lubricants, dyes, heavy metals, and fillers
The ultra-thin and consistent tube walls ensure that thermal transfer is precisely even along the surface of the tube. The unique cap design guarantees a perfect fit to prevent sample evaporation during thermal cycling. Tubes offer a maximum RCF of 4000 g. Tubes and caps are certified by lot number to be RNase-, DNase-, and pyrogen-free.
More Product Information