Hazardous Materials Shippers, Multi-Pack, UN Marked, Qorpak®
Supplier: Berlin Packaging
HMS-6454M HMS-6444M HMS-6442M HMS-6443M
16465-023CS 810.07 CAD
16465-023 16465-031 16465-033 16465-035
Hazardous Materials Shippers, Multi-Pack, UN Marked, Qorpak®
Mailers and Shippers
Four-pack multiple convenience packs suitable for shipping hazardous materials.
  • Plasticoated narrow mouth bottles for liquids
  • Plastic coated wide mouth bottles for solid and viscous materials
  • 120 mL shipper is not plastic coated
Certified packaging is supplied complete with safety coated glass container, Thermoset closure with foam backed fluoropolymer resin liner, corrugated outer packaging, cushioning material, absorbent, and tape. Shipper must not make any substitutions. Failure to heed this warning would invalidate package compliance.