Spectra/Por® 3 Dialysis Membranes, MWCO 3500, Spectrum® Laboratories
Supplier: Spectrum Laboratories
132720 132724 132725 S632724
25219-041EA 837.49 CAD
25219-041 25219-085 25219-121 28170-164 28170-166
Spectra/Por® 3 Dialysis Membranes, MWCO 3500, Spectrum® Laboratories
Tubing Dialysis Tubing
For dialysis and ultrafiltration in biochemical work.
Molecular weight cut-off is 3500. Pure, regenerated, natural cellulose. Free from fixed charges. Contain some glycerin and few impurities, which may be removed by washing. Porosity throughout roll is usually very uniform if roll is preserved carefully. Will change only slowly over a period of months if kept in the original polyethylene bag and refrigerated.
Roll length: 15.2 m (50')
Order Now
SPECIFICATIONS
Environmentally Preferable