Spectra/Por® 2 Dialysis Membranes, MWCO 12000 to 14000, Spectrum® Laboratories
Supplier: Spectrum Laboratories
132676 132678 132680 132682 132684
25218-402EA 1199.77 CAD
25218-402 25218-435 25218-468 25218-490 25218-548
Spectra/Por® 2 Dialysis Membranes, MWCO 12000 to 14000, Spectrum® Laboratories
Tubing Dialysis Tubing
Standard Regenerated Cellulose (RC) is composed of natural cellulose regenerated from cotton linters, therefore these membranes are perfectly suited for a broad range of laboratory dialysis.
This clear and flexible membrane type with a symmetric porosity is economical and sturdy, ideal for desalting, buffer exchanges, macromolecular purifications and other general applications. Possessing a broad range of chemical compatibilities, Standard RC membranes can be used with dilute strong acids and bases, concentrated weak acids and bases, most alcohols, and some mild or dilute organic solvents,More Product Information