Spectra/Por® Dialysis Tubing, Regenerated Cellulose, MWCO 12000 to 14000, Spectrum® Laboratories
Supplier: Spectrum Laboratories
S732700 S732706 S732709
25225-204CS 9191.43 CAD
25225-204 25225-226 25225-248 25225-260 25225-281
Spectra/Por® Dialysis Tubing, Regenerated Cellulose, MWCO 12000 to 14000, Spectrum® Laboratories
Tubing Dialysis Tubing
Standard dialysis tubing for all laboratory dialysis applications.
The tubing contains glycerin, water, and small amounts of sulfur compounds (approx. 0.1%).