Mark-M-Tubes Pre-Labeled Blood Typing Tubes, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
60A10BZ6 60A10BZ1 60A10BZ2 60A10BZ4 60B12BZ1 60B12BZ2 60B12BZ3 60B12BZ4 60B12BZ5 60B12BZ6
15404-412CS 456.36 CAD
15404-412 15404-402 15404-404 80080-468 89236-698 89236-700 89236-702 89236-704 89236-706 89236-708
Mark-M-Tubes Pre-Labeled Blood Typing Tubes, Kimble Chase
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
These culture tubes are manufactured with a fused ceramic paint to form writing labels that will not separate from the borosilicate glass tube. Labels measure 1.9W×3.5L cm (³/₄×1³/₈").
  • Pre-labeled with blood type
  • Convenient package quantities
  • Permanent labels placed to allow an unobstructed view of cell suspension
Certifications: Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements.