Insulated Shippers, Polyurethane, Sonoco ThermoSafe
Supplier: Sonoco Thermosafe
E238UPS E36UPS E89UPS E168UPS E327UPS E22UPS E90UPS E145 E124UPS E38 E90 E50UPS E186 E731 E731UPS E168 E438 E238 E337 E337UPS E186UPS E137 E137UPS E145UPS E200 E200UPS E89 E104 E104UPS E124 E126 E126UPS E65 E65UPS E174 E174UPS E69 E221 E38UPS E36 E21 E46 E46UPS E28 E50 E22 E15 E15UPS
14100-461EA 232.62 CAD
14100-461 14100-462 14100-464 14100-466 14100-468 14100-470 14100-472 73320-204 95054-984 82030-627 87004-421 89202-274 89401-704 89496-678 89496-680 89496-682 89496-684 89496-688 89496-690 89496-692 89496-696 89496-698 89496-700 89496-702 89496-704 89496-706 89496-708 89496-710 89496-712 89496-714 89496-716 89496-718 89496-720 89496-722 89496-724 89496-726 89496-728 89496-732 89496-736 89496-738 89496-740 89496-744 89496-746 89496-748 89496-750 89496-752 89496-754 89496-756
Insulated Shippers, Polyurethane, Sonoco ThermoSafe
Mailers and Shippers
ThermoSafe's high performance polyurethane insulated shipping containers meet the needs of the most demanding temperature-sensitive payloads.
  • Durable poly boxes
  • Protective shock absorption and cushioning
  • Ideal for temperature-sensitive items and long-range journeys
These poly shippers protect highly temperature-sensitive shipments from moderate and extreme ambient temperature exposure
Durable one-piece closed-cell polyurethane foam insulated shippers are CFC and HCFC-free and comply with international clean air regulations. Proprietary processMore Product Information