KONTES® NMR Sample Tubes, 10 mm, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
897320-0008 897320-0000
46600-764EA 143.29 CAD
46600-764 46600-766 46600-770 KT897320-0000 KT897325-0000 KT897335-0000
KONTES® NMR Sample Tubes, 10 mm, DWK Life Sciences
Tubes NMR Tubes
Designed for high resolution work with NMR spectrometers
Tubes are manufactured from ASTM Type 1 borosilicate glass under rigid quality control to meet exacting specifications for wall camber and variation. T.I.R. is the Total Indicator Reading. Each NMR tube is supplied with a polyethylene cap.
Order Now
SPECIFICATIONS
Environmentally Preferable