Dissolved Oxygen Probe, Beckman Coulter®
Supplier: Beckman Coulter
A57199 A57199 A58728 A58728
CABKA58728EA 172.34 CAD
BKA58728 BKA57199 CABKA58728 CABKA57199
Dissolved Oxygen Probe, Beckman Coulter®
Electrodes Dissolved Oxygen Electrodes
For use with Beckman Coulter® [PHI] series dissolved oxygen meters.
  • Integrated Temperature Probe
  • Integrated Cable
  • Locking, Waterproof Connector
  • Diameter: 16 mm
The probe features an integrated, one-meter cable with locking, waterproof connector.
Minimum sample flow: 5 cm/sec