Hazardous Material Shippers, HDPE, UN Marked, Qorpak®
Supplier: Berlin Packaging
HMS-6763M HMS-6765M HMS-6766M HMS-6767M
16465-043CS 746.22 CAD
16465-043 16465-047 16465-049 16465-051
Hazardous Material Shippers, HDPE, UN Marked, Qorpak®
Mailers and Shippers
Four-pack multiple convenience packs suitable for shipping hazardous materials.
  • Narrow mouth bottles for liquids
  • Wide mouth bottles for solid and viscous materials
Certified packaging is supplied complete with high density polyethylene container, polypropylene closure, corrugated outer packaging, cushioning material, absorbent, and tape.
Meet UN 4G requirements.
Warning: Shipper must not make any substitutions. Failure to heed this warning would invalidate package compliance.