HiVeg™ Peptones and Media, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
The HiVeg™ line of products consists of uniquely formulated, 100% vegetable-based peptones and media designed to work in conjunction with or to replace standard animal-based protein sources.
- 100% Vegetable Protein Source
- Eliminate the Risk of BSE/TSE
- Equivalent Performance with Animal-Based Peptones
The HiVeg™ line of products consists of uniquely formulated, 100% vegetable-based peptones and media designed to work in conjunction with or to replace standard animal-based protein sources. The products effectively eliminate BSE/TSE contamination concerns while providing results on par with animal-based products. The wide selection of peptones and media can meet the requirements of nearly any microbiology application.