VWR® Contour™ Top-Mounted Apron Legs
Supplier: VWR International
CFA-1159-A CFA-2159-A
CFA-1159-AEA 219.64 CAD
CFA-1159-A CFA-2159-A
VWR® Contour™ Top-Mounted Apron Legs
Furniture Casework
Each 5.1 cm (2") square leg is welded to 10.2 cm (4") square steel plate.
  • Contour casework is manufactured of prime grade, cold-rolled, U.S. Standard sheet steel
  • No ragged edges
  • Light neutral color
The complete apron is used to create a knee space for a desk area or to create a free-standing table. Apron components are sold separately.