Diagnostic Shippers and kPa Bags for Specimen Transport, Sonoco ThermoSafe
Supplier: Sonoco Thermosafe
470 473 472 476 570
16465-160CS 410.8 CAD
16465-160 16465-164 73320-122 73320-137 87003-906 87003-908 89049-960 89496-018
Diagnostic Shippers and kPa Bags for Specimen Transport, Sonoco ThermoSafe
Mailers and Shippers
ThermoSafe® Diagnostic Shippers are ready-to-use combination/triple packaging kits for specimen shipping.
  • Ready-to-use specimen mailers
  • Tested and certified to meet material and performance requirements for specimen transport
Small diagnostic specimen shipping system 87003-906 consists of a 95kPa plastic bag, two absorbent material sheets, a bubble wrap sheet, corrugated carton, and instruction sheet. Large diagnostic specimen shipping system 87003-908 consists of two 95kPa bags, four absorbent material sheets, two bubble wrap sheets, a corrugatedMore Product Information