VWR® Contour™ Standing Height Fume Hood Base Cabinet
Supplier: VWR International
CFG-2084-0F2 CFG-2084-A CFG-2085-A CFG-2086-A CFG-2088-A CFG-2089-A CWG-2084-A CWG-2085-A CWG-2086-A CWG-2088-A
CFG-2084-0F2EA 2125.73 CAD
CFG-2084-0F2 CFG-2084-A CFG-2085-A CFG-2086-A CFG-2088-A CFG-2089-A CWG-2084-A CWG-2085-A CWG-2086-A CWG-2088-A
VWR® Contour™ Standing Height Fume Hood Base Cabinet
Furniture Casework
Heavy-gauge steel full-frame construction.
  • Smooth, easy-to-maintain surfaces
  • Phosphate-coated with a baked, chemical-resistant, powder-coat finish
  • Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
A 2.54 cm (1") thick work surface creates an overall height of 91.4 cm (36"). All cupboard cabinets have one full-width adjustable shelf.