polyproLABS® Tall Storage Cabinets, Air Control
Supplier: Air Control
AC-300-3084 AC-300-3684 AC-300-4284 AC-300-4884 AC-310-3084 AC-310-3684 AC-310-4284 AC-310-4884
89008-726EA 13232.34 CAD
89008-726 89008-728 89008-730 89008-734 89008-736 89008-738 89008-740 89008-744
polyproLABS® Tall Storage Cabinets, Air Control
Furniture Casework
Corrosion-resistant casework cabinets are constructed of stress-relieved, acid-resistant, 1.3 cm (¹/₂") thick polypropylene.
  • No metals are used in cabinets
  • Toe space measures 10.2H×7.6D cm (4×3")
Joints are thermally welded by a special process that ensures leak-tight, flush unions. All hardware, including door hinges, screws, latches, drawer suspension systems, and handles, is constructed of polypropylene. Cabinets include one fixed center shelf and four adjustable shelves, made from solid 1.3 cm (¹/₂") thick polypropylene.