KoolTemp® Molded Polyurethane Containers, Cold Chain Technologies
Supplier: Cold Chain Technologies
U168-2-V U36-2-V U89-2-V U90-3-V
33500-416PK 1837.33 CAD
33500-416 33500-422 33500-424 33500-426
KoolTemp® Molded Polyurethane Containers, Cold Chain Technologies
Mailers and Shippers
Ultra-insulated containers maintain 2 to 8°C (35.6 to 46.4°F) in shipments over long time periods or in extreme weather conditions when used with refrigerant packs
Vapor-proof barrier enhances thermal effectiveness for a higher R-value (measure of thermal resistance) than other insulated containers. Containers are lightweight and one-piece molded. They come with an outer corrugate box sealed at the top with a soft foam polyurethane plug. Manufactured using a modular design that allows the user to recycle components. EPA-compliant, with no harmful CFCs. Meet or exceed the standards of all major shippers.