Nalgene® Petri Dish Rack, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
5922-0024 5921-0060 5920-0060
25405-104CS 669.5 CAD
25405-104 25405-159 25405-170
Nalgene® Petri Dish Rack, Thermo Scientific
Racks Petri Dish Racks
Designed for holding plastic disposable petri dishes, these polycarbonate racks provide full side visibility for checking cultures during incubation
Lightweight, easy to load, safe for transporting. Used for media preparation, inoculation, incubation, and storage.