Petri Dishes and Contact Dishes, Polystyrene, Greiner bio-one
Supplier: Greiner Bio-One
627102 627161 628102 628161 629161 629180 632102 632181 635102 639102 639161 664102 664161 688102 688161 632180 633180 633181
82050-536CS 276.13 CAD
82050-536 82050-540 82050-544 82050-548 82050-560 82050-562 82050-564 82050-572 82050-574 82050-578 82050-592 82050-596 82050-600 82050-914 82050-918 82051-066 82051-068 82050-566 82050-570 82050-584 82050-586
Petri Dishes and Contact Dishes, Polystyrene, Greiner bio-one
Culture Dishes Petri Dishes
These polystyrene petri dishes and contact dishes are available in different sizes and formats.
  • Available with or without vents
  • Nonsterile and sterile
  • Highly transparent polystyrene construction
Manufactured from highly transparent polystyrene, dishes are heat-resistant up to 60°C for use with hot agar.
Depending on the application, vented or non-vented petri dishes and contact dishes can be selected. If applicable, sterile dishes are also available.
Besides the standard round design, a square-profile petri dish is also available. Special modelsMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable