Casework, Laminate, Standing Height Base Cabinets, Cupboard and Drawer Cabinets
Supplier: Case Systems
B3100362436 B3100362448
33501-134EA 2210.07 CAD
33501-134 33501-136
Casework, Laminate, Standing Height Base Cabinets, Cupboard and Drawer Cabinets
Furniture Casework
Case Systems plastic laminate furniture combines modular and fixed laminate casework with coordinating hardware, trim, countertops, fixtures and many complementary accessories.
  • Key component of case systems casework is the plastic laminate
  • Case cabinetry gives adaptability necessary in today’s laboratories
A perfect choice for laboratory environments, plastic laminate provides superior resistance to chemicals, scratching, and abrasion. The radiused 3 mm PVC edging on doors and drawers further protects the casework from dents and marring whileMore Product Information