Casework, Laminate, Sitting Height Base Cabinets, Cupboard and Drawer Cabinets
Supplier: Case Systems
B3100302436 B3100302448
33501-130EA 2102.73 CAD
33501-130 33501-132
Casework, Laminate, Sitting Height Base Cabinets, Cupboard and Drawer Cabinets
Furniture Casework
Case systems plastic laminate furniture combines modular and fixed laminate casework with coordinating hardware, trim, countertops, fixtures and many complementary accessories.
  • The key component of case systems casework is the plastic laminate
  • Case cabinetry gives the adaptability necessary in today's laboratories
  • The result-cabinets that are as attractive as they are durable
A perfect choice for laboratory environments, plastic laminate provides superior resistance to chemicals, scratching, and abrasion. The radiused 3 mm PVC edging on doors and drawers further protects the casework from dents and marring while eliminating sharp edges and corners.
The high-quality laboratory grade plastic laminate cabinets are manufactured to exact tolerances with consistent quality standards. Construction features an industrial-grade, environmentally responsible WOODSTALK® engineered board core from Dow BioProducts, using formaldehyde-free resin to bind wheat-straw fiber, and heavy-duty institutional grade hardware.
The unique hardware system provides rigid support for vertical integration without limiting adjustability. Vertical supports are slotted, allowing infinite height adjustment of overhead cabinets, electrical raceways, services, and reagent shelves.
Cupboard cabinet has one shelf, two doors, and two drawers. Sitting height base cabinets are common for traditional laboratory and technical environments. They are usually installed for permanent or semi-permanent layouts. Color: Beige.
Ordering information: Case systems laminate laboratory furniture has all the components necessary for a complete laboratory set-up. For further information, contact your Avantor Furniture Specialist.