Nunc® Bio-Assay Dishes, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
240835 240845
CA25384-002CS 527.1 CAD
CA25384-002 CA62409-456
Nunc® Bio-Assay Dishes, Sterile, Thermo Scientific
Culture Dishes Petri Dishes
These clear, polystyrene dishes feature a large growth area and are suitable for performing agar diffusion assays, screening large numbers of colonies in cloning experiments, and for culturing of bacteria and fungi
The low-profile dish is specifically designed for HTS genomic screening and automated colony picking. Compatible with robotics and instrumentation, the dish features stacking ribs and orientation notches. Low height saves space.
They can also be used as a moisture chamber for incubation of NUNC® MicroWell™ plates. When lined with wet filter paper, the dishes can also serveMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable