Nunc® UpCell® MultiDishes, Thermo Scientific
Supplier: Thermo Fisher Scientific
174905 174906 174897 174898 174899 174900 174901 174903 174902 174904
CA89089-598CS 1278.27 CAD
CA89089-598 CA89089-600 CA89089-602 CA89089-604 CA89089-606 CA89089-608 CA89089-610 CA89089-614 CA89089-612 CA89089-616
Nunc® UpCell® MultiDishes, Thermo Scientific
Culture Dishes Cell Culture Dishes
Single-use vented multidishes feature a temperature-responsive cell culture surface that is ideal for culture passaging, single-cell analyses, and cell transplantation research
The surface allows harvesting of cells with intact surface proteins. Adherent cells are released by reducing the temperature of the cell culture. High cell viability is achieved with minimal hands-on time and without trypsinization or physical force. Ideal for harvesting of cell sheets and creations of 3-D tissue models held together by normal cell junctions and the extracellular matrix. Sterile.