CELLSTAR® Tissue Culture Dishes, Polystyrene, Sterile, Greiner Bio-One
Supplier: Greiner Bio-One
639160 627160 628160 664160 628160-TRI 639160-TRI 664160-TRI
82050-598CS 250.38 CAD
82050-598 82050-538 82050-546 82050-916 75799-854 75799-856 75799-858
CELLSTAR® Tissue Culture Dishes, Polystyrene, Sterile, Greiner Bio-One
Culture Dishes Cell Culture Dishes
CELLSTAR® dishes are manufactured from high-quality virgin polystyrene and are treated for optimal cell attachment and proliferation.
  • High-quality virgin polystyrene construction
  • Nonpyrogenic
  • Free of RNase and DNase
  • Stackable
  • Uniform gas exchange and reduced condensation
Dishes offer distortion-free clarity. They are nonpyrogenic and RNase- and DNase-free. All dishes feature stacking rims for simple storage and ease in handling. Vented versions provide uniform gas exchange and reduced condensation.