Revco® Accessories for Refrigerators and Freezers, Thermo Scientific
Supplier: Thermo Fisher Scientific
6041 6704 6703 6702 6701 6700 6694 6383-7D 6903 6206 6224TA 4706 CA14228-372 6736 6383-5 6202 6383-7A 90520H11 6183-7 90520H07 6105
14228-255EA 366.05 CAD
14228-255 14228-308 14228-310 14228-312 14228-314 14228-318 14228-352 14228-378 14228-512 55701-740 55702-851 89176-758 CA14228-372 14228-356 14228-374 73620-026 73620-050 73650-100 CA55702-630 14228-372 55702-641 CA89369-376
Revco® Accessories for Refrigerators and Freezers, Thermo Scientific
Refrigerators
Temperature recorders use pressure-sensitive chart paper (sold separately) and do not require inking
  • A built-in surge suppressor is standard on Ultima II series models
  • A freestanding surge suppressor is available for Elite® and Value series models
  • Revco® ultra-low temperature freezers include one 2.5 cm (1") diameter pass-thru port
  • Additional ports are available to provide openings for probes, cables, or leads
The remote alarm module in temperature recorder provides an audible and visual signal in the event of a temperature rise or power failure