Nalgene® Oak Ridge Centrifuge Tubes, Polypropylene Copolymer, Thermo Scientific
Supplier: Thermo Fisher Scientific
3119-0030 3119-0028 3119-0050 3119-0010 3139-0010 3139-0016 3139-0030 3139-0050
21009-364CS 1980.21 CAD
21009-364 21009-375 21009-386 21009-397 21010-523 21010-524 21010-567 21010-578
Nalgene® Oak Ridge Centrifuge Tubes, Polypropylene Copolymer, Thermo Scientific
Tubes Centrifuge Tubes
Contact-clear, leakproof polypropylene copolymer tubes allow easy examination of contents.
  • Repeatedly autoclavable
  • Can be used upto 50000 ×g in refrigerated centrifuges
Greater chemical resistance than polycarbonate tubes. Before autoclaving, place closure on top of container without engaging threads.
Certifications: Meets OSHA Standard 29CFR, Part 1910.1030 for use as protection against bloodborne pathogens.
Ordering information: Tubes with sealing caps are supplied with polypropylene screw closure and retained silicone gasket. Tubes with screwMore Product Information